Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF86300445

Iron

rod, 4.0 mm diameter, length 200 mm, high purity 99.99+%

Synonym(s):

FE007928

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$861.00

$861.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$861.00

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

$861.00


Check Cart for Availability

Request a Bulk Order

assay

≥99.99%

form

rod

manufacturer/tradename

Goodfellow 863-004-45

resistivity

9.71 μΩ-cm

L

200 mm

diameter

4.0 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service