GF86267751
Polymethylmethacrylate
(PMMA, ACRYLIC), sheet, thickness 0.38 mm, size 500 × 600 mm, condition impact modified
Synonym(s):
ME303001
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352100
form
sheet
manufacturer/tradename
Goodfellow 862-677-51
size
500 × 600 mm
thickness
0.38 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service