Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF81988464

Nickel

wire reel, 100m, diameter 0.05mm, as drawn, 99%

Synonym(s):

Nickel, NI005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23
Product GF81988464 is not currently sold in your country. Contact Technical Service

assay

99%

form

wire

manufacturer/tradename

Goodfellow 819-884-64

resistivity

6.97 μΩ-cm, 20°C

L × diam.

100 m × 0.05 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meinar Nur Ashrin et al.
Journal of immunology (Baltimore, Md. : 1950), 192(9), 4025-4031 (2014-03-29)
Ni is the most frequent cause of contact allergy induced by metals. However, the underlying mechanism of this induction is unknown. Our previous research demonstrates that activation of dendritic cells (DCs) through p38MAPK/MKK6 is required for Ni-induced allergy in mice.
Erkai He et al.
Environmental pollution (Barking, Essex : 1987), 188, 17-26 (2014-02-18)
Protons and other cations may inhibit metal uptake and alleviate metal toxicity in aquatic organisms, but less is known about these interactions in soil organisms. The present study investigated the influence of solution chemistry on uptake and toxicity of Ni
Filaggrin is a predominant member of the denaturation-resistant nickel-binding proteome of human epidermis.
Katrine Ross-Hansen et al.
The Journal of investigative dermatology, 134(4), 1164-1166 (2013-10-26)
Sébastien Hulo et al.
Toxicology letters, 226(1), 63-69 (2014-02-11)
To evaluate manganese in exhaled breath condensate (Mn-EBC) as an indicator of exposure to fumes from metal inert gas welding process. We collected EBC and urine from 17 welders and 16 unexposed control subjects after 5 days exposure. Concentrations of
Som Niyogi et al.
Aquatic toxicology (Amsterdam, Netherlands), 150, 36-44 (2014-03-19)
The freshwater pulmonate snail, Lymnaea stagnalis, is the most sensitive aquatic organism tested to date for Ni. We undertook a series of experiments to investigate the underlying mechanism(s) for this observed hypersensitivity. Consistent with previous experiments, juvenile snail growth in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service