Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF81811650

T1 - thermocouple alloy, Ni90Cr10

insulated wire, 100m length, 0.05mm conductor diameter, 0.005mm insulation thickness, polyurethane insulation

Synonym(s):

NI095821

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23
Product GF81811650 is not currently sold in your country. Contact Technical Service

description

polyurethane insulation
thermocouple wire

form

wire (insulated)

manufacturer/tradename

Goodfellow 818-116-50

insulation thickness

0.005 mm

wire L × diam.

100 m × 0.05 mm , conductor diameter

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Application

may be used as one component of a thermocouple

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service