GF81809589
Copper-beryllium alloy, Cu98Be2
foil, 150 x 150mm, 0.05mm thickness, half hard, light tight (lt)
Synonym(s):
CU070300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
31152304
NACRES:
NA.23
description
light tight (lt)
form
foil (half hard)
manufacturer/tradename
Goodfellow 818-095-89
size × thickness
150 mm × 150 mm × 0.05 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service