Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF81191998

Vanadium

rod, 3.5 mm diameter, length 100 mm, purity 99.8%

Synonym(s):

V 007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

assay

99.8%

form

rod

manufacturer/tradename

Goodfellow 811-919-98

resistivity

24.8-26.0 μΩ-cm, 20°C

L

100 mm

diameter

3.5 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service