GF81063929
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1
foil, 25 x 25mm, 0.007mm thickness, as rolled, light tight (lt)
Synonym(s):
FE010240
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
description
light tight (lt)
form
foil (as rolled)
manufacturer/tradename
Goodfellow 810-639-29
size × thickness
25 mm × 25 mm × 0.007 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service