Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF79914268

Copper-beryllium alloy, Cu98Be2

foil, 10m coil, .3mm coil width, 0.15mm thickness, half hard

Synonym(s):

CU070361

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23
Product GF79914268 is not currently sold in your country. Contact Technical Service

description

temper (half hard)

form

foil (coil)

manufacturer/tradename

Goodfellow 799-142-68

L × width × thickness

10 m × 0.3 mm × 0.15 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service