Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF79790709

Gallium

lump, 25 mm max. lump size, weight 50 g, purity 99.99%

Synonym(s):

GA006120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141715
NACRES:
NA.23

assay

99.99%

form

lumps

manufacturer/tradename

Goodfellow 797-907-09

weight

50 g

max. lump size

25 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service