Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF79168674

Hastelloy® B - corrosion resistant alloy, Ni62Mo28Fe5CrMnSi

foil, 25 x 25mm, 0.010mm thickness, light tight (lt)

Synonym(s):

NI010250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Product GF79168674 is not currently sold in your country. Contact Technical Service

form

foil (light tight (lt))

manufacturer/tradename

Goodfellow 791-686-74

size × thickness

25 x 25 mm × 0.010 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Hastelloy is a registered trademark of Haynes International, Inc.
Hastelloy is a registered trademark of Haynes International, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service