Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF78741603

Aluminum-magnesium alloy, Al97Mg3

1000mm tube, 1.5mm outside diameter, 0.125mm wall thickness, 1.25mm inside diameter, as drawn

Synonym(s):

AL017250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23
Product GF78741603 is not currently sold in your country. Contact Technical Service

form

tubes

manufacturer/tradename

Goodfellow 787-416-03

L × wall thickness

1000 mm × 0.125 mm

O.D. × I.D.

1.5 mm × 1.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service