Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF78356863

Stainless Steel - AISI 304 alloy, FeCr18Ni10

100m wire, 0.075mm diameter, annealed

Synonym(s):

FE225120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23
Product GF78356863 is not currently sold in your country. Contact Technical Service

form

wire

manufacturer/tradename

Goodfellow 783-568-63

L × diam.

100 m × 0.075 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service