Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF78170003

Indium

wire reel, 2m, diameter 0.75mm, as drawn, 99.999%

Synonym(s):

Indium, IN005125

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

assay

99.999%

form

wire

manufacturer/tradename

Goodfellow 781-700-03

resistivity

8.37 μΩ-cm

L × diam.

2 m × 0.75 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Schneider et al.
Accounts of chemical research, 45(8), 1331-1344 (2012-05-26)
The development of innovative metal catalysis for selective bond formation is an important task in organic chemistry. The group 13 metal indium is appealing for catalysis because indium-based reagents are minimally toxic, selective, and tolerant toward various functional groups. Among
New generation of palladium-indium-silver dental cast alloys: a review.
M L Malhotra
Trends & techniques in the contemporary dental laboratory, 9(4), 65-68 (1992-05-01)
Methodologic and basic aspects of indium-111 platelets.
J H Joist et al.
Seminars in thrombosis and hemostasis, 9(2), 86-99 (1983-04-01)
Alex N Eberle et al.
Journal of molecular recognition : JMR, 16(5), 248-254 (2003-10-03)
Following the first synthesis of tritiated alpha-melanocyte-stimulating hormone (alpha-MSH, alpha-melanotropin) in 1974 by Medzihradszky et al., several alpha-MSH analogs were designed containing between 2 and 12 tritium atoms, the latter of which displayed a specific radioactivity of 12.21 GBq/micromol (330
Mikio Nakajima et al.
Congenital anomalies, 48(4), 145-150 (2008-11-06)
Indium, a precious metal classified in group 13 (IIIB) in the periodic table, has been used increasingly in the semiconductor industry. Because indium is a rare metal, technology for indium recycling from transparent conducting films for liquid crystal displays is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service