GF77513946
Magnesium
rod, 50 mm diameter, length 100 mm, purity 99.9%
Synonym(s):
MG007950
About This Item
vapor pressure
1 mmHg ( 621 °C)
assay
99.9%
form
rod
autoignition temp.
950 °F
manufacturer/tradename
Goodfellow 775-139-46
resistivity
4.46 μΩ-cm, 20°C
L
100 mm
diameter
50 mm
bp
1090 °C (lit.)
mp
648 °C (lit.)
density
1.74 g/mL at 25 °C (lit.)
SMILES string
[Mg]
InChI
1S/Mg
InChI key
FYYHWMGAXLPEAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service