Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF77111108

Tantalum

mesh, 150x150mm, nominal aperture 0.43mm, wire diameter 0.075mm, 50x50 wires/inch, open area 72%, plain weave mesh

Synonym(s):

Tantalum, TA008710, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

form

mesh

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 771-111-08

resistivity

13.5 μΩ-cm, 20°C

size × nominal aperture × diam.

150 x 150 mm × 0.43 mm × 0.075 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett Levine et al.
The journal of knee surgery, 20(3), 185-194 (2007-08-02)
Porous tantalum represents an alternative metal for primary and revision total knee arthroplasty (TKA) with several unique properties. Tantalum is a transition metal, which in its bulk form has shown excellent biocompatibility and is safe to use in vivo as
Nilesh Patil et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 89(1), 242-251 (2008-10-08)
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone.
J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Juncai Deng et al.
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi, 28(2), 419-422 (2011-05-25)
This paper describes the biological characteristics of the metal tantalum (Ts) and the application status of tantalum in artificial joints, bone necrosis, spine, defects of bone and other aspects of bone. The early clinical application results of tantalum in bone
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service