Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF76957004

Vanadium

microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%

Synonym(s):

Vanadium, V 004600

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

assay

99.8%

form

foil

manufacturer/tradename

Goodfellow 769-570-04

resistivity

24.8-26.0 μΩ-cm, 20°C

diam. × thickness

25 mm × 0.25 μm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lise Coderre et al.
Canadian journal of physiology and pharmacology, 82(10), 833-839 (2004-12-02)
Inorganic and organic compounds of vanadium have been shown to exhibit a large range of insulinomimetic effects in the cardiovascular system, including stimulation of glucose transporter 4 (GLUT-4) translocation and glucose transport in adult cardiomyocytes. Furthermore, administration of vanadium compounds
Maja A Larsson et al.
Journal of hazardous materials, 296, 158-165 (2015-04-29)
Blast furnace (BF) slags are commonly applied as soil amendments and in road fill material. In Sweden they are also naturally high in vanadium. The aim of this study was to assess the vanadium bioavailability in BF slags when applied
D M Smith et al.
QJM : monthly journal of the Association of Physicians, 101(5), 351-358 (2008-03-06)
To assess the effectiveness of oral vanadium supplementation for glycaemic control in type 2 diabetes by conducting a systematic review of the literature. Eligible studies were identified by searching 14 databases using standardized terms. Experts, study authors and manufacturers were
Dieter Rehder
Journal of inorganic biochemistry, 102(5-6), 1152-1158 (2008-02-08)
In the present focused review, vanadate-dependent haloperoxidases and vanadate-inhibited enzymes which catalyze the hydrolysis of phosphoester bonds are addressed. In these systems, vanadate [HxVO4](3-x)(-) is covalently coordinated to the imidazolyl moiety of an active site histidine, with a geometrical arrangement
Birgit Schwenzer et al.
ChemSusChem, 4(10), 1388-1406 (2011-11-22)
Large-scale energy storage has become the main bottleneck for increasing the percentage of renewable energy in our electricity grids. Redox flow batteries are considered to be among the best options for electricity storage in the megawatt range and large demonstration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service