Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF76896400

Chromium

foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.99+%

Synonym(s):

Chromium, CR000040

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
MDL number:
UNSPSC Code:
12141709
PubChem Substance ID:
NACRES:
NA.23

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 768-964-00

resistivity

12.7 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.0025 mm

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

Inchi Key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Dhal et al.
Journal of hazardous materials, 250-251, 272-291 (2013-03-08)
Chromium is a highly toxic non-essential metal for microorganisms and plants, and its occurrence is rare in nature. Lower to higher chromium containing effluents and solid wastes released by activities such as mining, metal plating, wood preservation, ink manufacture, dyes
A M Urbano et al.
Current drug metabolism, 13(3), 284-305 (2012-03-30)
For over a century, chromium (Cr) has found widespread industrial and commercial use, namely as a pigment, in the production of stainless steel and in chrome plating. The adverse health effects to the skin and respiratory tract of prolonged exposure
Sandra S Wise et al.
Mutation research, 733(1-2), 78-82 (2011-12-24)
Many metals serve as micronutrients which protect against genomic instability. Chromium is most abundant in its trivalent and hexavalent forms. Trivalent chromium has historically been considered an essential element, though recent data indicate that while it can have pharmacological effects
Akiko Maehara et al.
EuroIntervention : journal of EuroPCR in collaboration with the Working Group on Interventional Cardiology of the European Society of Cardiology, 10(4), 475-483 (2014-08-21)
To report serial intravascular ultrasound (IVUS) findings of bifurcation lesions treated with the dedicated Tryton Side Branch Stent to assess mechanisms of restenosis. The Tryton FIM study was a multicentre, prospective, single-arm, "first-in-man" study to treat de novo bifurcation lesions.
Bo Xia et al.
Toxicology letters, 228(3), 241-247 (2014-05-27)
Chromium is a potent human mutagen and carcinogen. The capability of chromium to cause cancers has been known for more than a century, and numerous epidemiological studies have been performed to determine its carcinogenicity. In the post-genome era, cancer has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service