Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF76141829

Tin

wire reel, 20m, diameter 0.25mm, as drawn, 99.99+%

Synonym(s):

Tin, SN005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Product GF76141829 is not currently sold in your country. Contact Technical Service

assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 761-418-29

resistivity

11 μΩ-cm, 20°C

L × diam.

20 m × 0.25 mm

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Tin in the environment].
Iu P Popov et al.
Gigiena i sanitariia, (9)(9), 55-57 (1983-09-01)
Inorganic tin: chemistry, disposition and role in nuclear medicine diagnostic skeletal imaging agents.
M D Francis et al.
International journal of nuclear medicine and biology, 8(2-3), 145-152 (1981-01-01)
The newer essential trace elements, chromium, tin, nickel, vanadium and silicon.
W Mertz
The Proceedings of the Nutrition Society, 33(3), 307-313 (1974-12-01)
Y Arakawa
Sangyo eiseigaku zasshi = Journal of occupational health, 39(1), 1-20 (1997-01-01)
Tin generates a wide variety of biological activities deriving from its chemical character. In this article, the biological activities of tin compounds are reviewed with a focus on the connection with immunity. The table of contents is as follows: Introduction
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service