GF76128958
Phosphor bronze alloy, Cu94Sn6
foil, 2m coil, 20mm coil width, 0.4mm thickness, hard
Synonym(s):
CU050280
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
description
temper (hard)
form
foil (coil)
manufacturer/tradename
Goodfellow 761-289-58
L × W × thickness
2 m × 20 mm × 0.4 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service