Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF76110476

Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1

100g lump, 50mm max. lump size, pellets

Synonym(s):

FE086100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Product GF76110476 is not currently sold in your country. Contact Technical Service

form

pellets

manufacturer/tradename

Goodfellow 761-104-76

weight

100 g

max. lump size

50 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service