Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF75799647

Dural® - aluminum-copper-magnesium alloy, Al95Cu4Mg1

1000mm tube, 0.81mm outside diameter, 0.051mm wall thickness, 0.708mm inside diameter, as drawn

Synonym(s):

AL047110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 757-996-47

L × wall thickness

1000 mm × 0.051 mm

O.D. × I.D.

0.81 mm × 0.708 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Dural is a registered trademark of Lonza-Werke GmbH
Dural is a registered trademark of Lonza-Werke GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service