Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF75525341

Molybdenum

powder, 350 max. part. size (micron), weight 1000 g, purity 99.9+%

Synonym(s):

MO006011

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23
Product GF75525341 is not currently sold in your country. Contact Technical Service

assay

≥99.9%

form

powder

manufacturer/tradename

Goodfellow 755-253-41

resistivity

5.0 μΩ-cm, 20°C

weight

1000 g

max. part. size

350 (micron)

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Flam. Sol. 1 - Repr. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service