Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF75524733

Monel® alloy 400, Ni65Cu33Fe2

1000mm tube, 1.9mm outside diameter, 0.275mm wall thickness, 1.35mm inside diameter, as drawn

Synonym(s):

NI117300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Product GF75524733 is not currently sold in your country. Contact Technical Service

form

tubes

manufacturer/tradename

Goodfellow 755-247-33

L × wall thickness

1000 mm × 0.275 mm

O.D. × I.D.

1.9 mm × 1.35 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service