Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF74984473

Thallium

rod, 250mm, diameter 8.0mm, extruded, 99.99+%

Synonym(s):

Thallium, TL007915

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Tl
CAS Number:
Molecular Weight:
204.38
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.99%

form

rod

manufacturer/tradename

Goodfellow 749-844-73

resistivity

18 μΩ-cm, 20°C

L × diam.

250 mm × 8.0 mm

bp

1457 °C (lit.)

mp

303 °C (lit.)

SMILES string

[Tl]

InChI

1S/Tl

InChI key

BKVIYDNLLOSFOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Cvjetko et al.
Arhiv za higijenu rada i toksikologiju, 61(1), 111-119 (2010-03-27)
Thallium is a naturally occurring trace element, widely distributed in the earth's crust, but at very low concentrations. It does not have a known biological use and does not appear to be an essential element for life. It has been
Qihang Wu et al.
Environmental science and pollution research international, 22(15), 11478-11487 (2015-03-31)
Thallium (Tl) pollution in agricultural areas can pose hidden danger to humans, as food consumption is the key exposure pathway of Tl. Owing to the extreme toxicity of Tl, removal of Tl from soil becomes necessary to minimize the Tl-related
S Galván-Arzate et al.
Toxicology letters, 99(1), 1-13 (1998-11-04)
Thallium (T1+) is a toxic heavy metal which was accidentally discovered by Sir William Crookes in 1861 by burning the dust from a sulfuric acid industrial plant. He observed a bright green spectral band that quickly disappeared. Crookes named the
R S Hoffman
Journal of toxicology. Clinical toxicology, 38(7), 767-775 (2001-02-24)
Thallium salts were once used to treat a variety of disorders such as tuberculosis, sexually transmitted diseases, and ringworm of the scalp. Although the clinical manifestations and treatment of patients with thallium poisoning are well characterized, little information is available
Tangfu Xiao et al.
The Science of the total environment, 421-422, 51-58 (2011-04-26)
It is well known that thallium (Tl) is a non-essential and toxic metal to human health, but less is known about the geo-environmentally-induced Tl pollution and its associated health impacts. High concentrations of Tl that are primarily associated with the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service