Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF74404751

Gadolinium

foil, thickness 0.25 mm, size 50 × 50 mm, purity 99%

Synonym(s):

GD000250

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,490.00

$1,490.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,490.00

About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

$1,490.00


Check Cart for Availability

Request a Bulk Order

assay

99%

form

foil

manufacturer/tradename

Goodfellow 744-047-51

resistivity

126 μΩ-cm, 20°C

size

50 × 50 mm

thickness

0.25 mm

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Water-react 3

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service