Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF73974954

Copper - O.F.H.C.

foil, 50x50mm, thickness 0.125mm, hard, 99.95+%

Synonym(s):

Copper, CV000590

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
12141711
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 739-749-54

resistivity

1.673 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.125 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donita C Brady et al.
Nature, 509(7501), 492-496 (2014-04-11)
The BRAF kinase is mutated, typically Val 600→Glu (V600E), to induce an active oncogenic state in a large fraction of melanomas, thyroid cancers, hairy cell leukaemias and, to a smaller extent, a wide spectrum of other cancers. BRAF(V600E) phosphorylates and
Hiroki Serizawa et al.
Organic letters, 16(13), 3456-3459 (2014-06-14)
The direct synthesis of pentafluoroethyl copper (CuC2F5) from a cuprate reagent and ethyl pentafluoropropionate as one of the most economical and useful pentafluoroethyl sources was accomplished. The advantages of this method are; all the reagents employed are low-cost and operationally
Jing Zheng et al.
Organic letters, 16(13), 3560-3563 (2014-06-25)
A Rh(III)-catalyzed selective coupling of N-methoxy-1H-indole-1-carboxamide and aryl boronic acids is reported. The coupling is mild and efficient toward diverse product formation, with selective C-C and C-C/C-N bond formation. Kinetic isotope effects studies were conducted to reveal a mechanism of
Lelita T Braiterman et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), E1364-E1373 (2014-04-08)
Wilson disease (WD) is a monogenic autosomal-recessive disorder of copper accumulation that leads to liver failure and/or neurological deficits. WD is caused by mutations in ATP7B, a transporter that loads Cu(I) onto newly synthesized cupro-enzymes in the trans-Golgi network (TGN)
Nathalie Adam et al.
Environmental pollution (Barking, Essex : 1987), 194, 130-137 (2014-08-12)
In this study the uptake of ZnO and CuO nanoparticles by Daphnia magna was tested. Daphnids were exposed during 48 h to acute concentrations of the nanoparticles and corresponding metal salts. The Daphnia zinc and copper concentration was measured and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service