Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF71418416

Nickel

tube, 200mm, outside diameter 1.57mm, inside diameter 0.63mm, wall thickness 0.47mm, hard, 99.5%

Synonym(s):

Nickel, NI007260

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

99.5%

form

tubes

manufacturer/tradename

Goodfellow 714-184-16

resistivity

6.97 μΩ-cm, 20°C

L × O.D. × I.D.

200 mm × 1.57 mm × 0.63 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Organic letters, 16(7), 1904-1907 (2014-03-29)
A general method for the alkenylation of alkyl electrophiles using nearly stoichiometric amounts of the air- and moisture-stable potassium organotrifluoroborates has been developed. Various functional groups were tolerated on both the nucleophilic and electrophilic partner. Reactions of highly substituted E-
Erkai He et al.
Environmental pollution (Barking, Essex : 1987), 188, 17-26 (2014-02-18)
Protons and other cations may inhibit metal uptake and alleviate metal toxicity in aquatic organisms, but less is known about these interactions in soil organisms. The present study investigated the influence of solution chemistry on uptake and toxicity of Ni
Filaggrin is a predominant member of the denaturation-resistant nickel-binding proteome of human epidermis.
Katrine Ross-Hansen et al.
The Journal of investigative dermatology, 134(4), 1164-1166 (2013-10-26)
Hao Yuan et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(4), 764-772 (2014-03-19)
In surgery-first accelerated orthognathic surgery, the clinical phenomenon of accelerated orthodontic tooth movement after osteotomy is a benefit compared with the conventional approach. However, because much of the literature on this phenomenon is based on empirical evidence and case reports
Crystal Cox et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 505-513 (2014-04-08)
Nickel-titanium closed-coil springs are purported to deliver constant forces over extended ranges of activation and working times. In-vivo studies supporting this claim are limited. The objective of this study was to evaluate changes in force-decay properties of nickel-titanium closed-coil springs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service