Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF69018082

Zinc

foil, 300x300mm, thickness 0.125mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000270

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Product GF69018082 is not currently sold in your country. Contact Technical Service

vapor pressure

1 mmHg ( 487 °C)

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 690-180-82

resistivity

5.8 μΩ-cm, 20°C

size × thickness

300x300 mm × 0.125 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengquan Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(22), 5823-5828 (2004-10-09)
A solution surface-erosion route was successfully employed to produce one-dimensional (1D) ZnO nanostructures. ZnO nanorod arrays and three-dimensional urchin-like assemblies could be selectively obtained with different manipulations. In this process, zinc foil was introduced to an organic solution system and
Yang Liu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(23), 6667-6673 (2007-05-31)
Well-aligned zinc oxide microrod and microtube arrays with high aspect ratios were fabricated on zinc foil by a simple solution-phase approach in an aqueous solution of ethylenediamine (en). The shape of the ZnO microstructures can be easily modulated from rods
Huibo Chen et al.
Nanoscale research letters, 5(3), 570-575 (2009-01-01)
We reported the optical and wettability properties of aligned zinc oxide micro/nanotube arrays, which were synthesized on zinc foil via a simple hydrothermal method. As-synthesized ZnO micro/nanotubes have uniform growth directions along the [0001] orientations with diameters in the range
Pritesh Hiralal et al.
ACS nano, 4(5), 2730-2734 (2010-04-27)
Solid-state and flexible zinc carbon (or Leclanche) batteries are fabricated using a combination of functional nanostructured materials for optimum performance. Flexible carbon nanofiber mats obtained by electrospinning are used as a current collector and cathode support for the batteries. The
A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service