GF66508184
Cellulose Acetate
(CA), film, thickness 0.035 mm, L 10 m, natural
Synonym(s):
AC311035
Select a Size
$624.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$624.00
Notify Me
Get notified when this item is ready to ship via email.
Recommended Products
form
film
manufacturer/tradename
Goodfellow 665-081-84
L
10 m
thickness
0.035 mm
color
natural
Related Categories
General description
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service