GF65516801
Inconel® alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi
foil, 10mm disks, 0.25mm thickness, annealed
Synonym(s):
NI130300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
form
annealed
foil (disks)
manufacturer/tradename
Goodfellow 655-168-01
size × thickness
10 mm × 0.25 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service