GF65371271
Carbon
lump, 1.5 mm max. lump size, weight 50 g, purity 99.5%
Synonym(s):
Activated charcoal, C 006101, Charcoal activated
About This Item
vapor pressure
<0.1 mmHg ( 20 °C)
assay
99.5%
form
lumps
autoignition temp.
842 °F
manufacturer/tradename
Goodfellow 653-712-71
resistivity
1375 μΩ-cm, 20°C (graphite)
weight
50 g
max. lump size
1.5 mm
mp
3550 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service