Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF61330452

Platinum

wire reel, 2m, diameter 0.20mm, annealed, may be used as one component of a thermocouple, 99.99%

Synonym(s):

Platinum, PT005130

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$679.00

$679.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$679.00

About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

$679.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 613-304-52

resistivity

10.6 μΩ-cm, 20°C

L × diam.

2 m × 0.20 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

Inchi Key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Iafisco et al.
Journal of inorganic biochemistry, 117, 237-247 (2012-07-25)
The present review focuses on the "drug targeting and delivery" approach of the selective transportation of cisplatin to bone tumors and bone metastases. This aim is realized by binding cisplatin to (bis)phosphonate ligands or their derivatives. Geminal bisphosphonates are in
Himashinie V K Diyabalanage et al.
Cancer letters, 329(1), 1-8 (2012-10-04)
One of the most promising strategies to increase the efficacy of standard chemotherapy drugs is by combining them with low doses of histone deacetylases inhibitors (HDACis). Regarded as chemosensitizers, the addition of well-tolerated doses of HDACis to platinum-based chemotherapeutics has
Fausto Petrelli et al.
Lung cancer (Amsterdam, Netherlands), 81(3), 337-342 (2013-07-31)
The sole agents pemetrexed (PEM), docetaxel and anti-EGFR agents are approved second-line therapies for non-small cell lung cancer (NSCLC) after failure with cisplatin-based doublets. The potential usefulness of platinum-based doublets as rechallenge for second-line chemotherapy has not yet been established.
Thorsten Langer et al.
Trends in pharmacological sciences, 34(8), 458-469 (2013-06-19)
Childhood cancer survival rates are now nearly 80% in more developed European countries because of improved therapies and better supportive care. Platinum chemotherapy drugs, such as cisplatin and carboplatin, are the cornerstone of many effective therapeutic protocols for childhood cancer.
Nicoletta Colombo
Future oncology (London, England), 9(12 Suppl), 19-23 (2013-11-15)
Ovarian cancer is the leading cause of gynecological cancer deaths worldwide. Despite primary treatment with platinum-containing regimens, the majority of women will experience recurrent disease and subsequent death. Recurrent ovarian cancer remains a challenge for successful management, and the choice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service