Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF61122428

Hafnium

foil, 15mm disks, thickness 0.025mm, annealed, 97%

Synonym(s):

Hafnium, HF000200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
MDL number:
UNSPSC Code:
12141718
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

97%

form

foil

manufacturer/tradename

Goodfellow 611-224-28

resistivity

29.6 μΩ-cm, 0°C

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service