Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF59674977

Inconel® X750 - heat resisting alloy, Ni74Cr15Fe7TiAlNb

foil, 200 x 200mm, 0.028mm thickness, as rolled, light tight (lt)

Synonym(s):

NI160220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Pricing and availability is not currently available.

description

light tight (lt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 596-749-77

size × thickness

200 mm × 200 mm × 0.028 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service