Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF59427513

Holmium

wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%

Synonym(s):

Holmium, HO005105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 594-275-13

L × diam.

200 mm × 0.5 mm

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A systematic review of the clinical efficacy and effectiveness of the holmium:YAG laser in urology.
I Larizgoitia et al.
BJU international, 84(1), 1-9 (1999-08-12)
Sean Pierre et al.
World journal of urology, 25(3), 235-239 (2007-03-07)
The efficiency and safety profile of the holmium laser has made this tool a versatile multi-purpose instrument for use in the endoscopic treatment of a wide variety of urologic disorders, in particular urinary calculi. Herein we review holmium laser physics
Benoit Molimard et al.
Urology, 76(6), 1334-1337 (2010-06-25)
To assess the outcome of flexible ureterorenoscopy (F-URS) with the holmium laser in treating stones in the horseshoe kidney (HSK). We retrospectively reviewed the records of 17 patients with a HSK stone (17 renal units) who had undergone F-URS with
A Le Duc et al.
European urology, 35(2), 155-160 (1999-02-06)
This review of holmium laser resection of the prostate (HoLRP) summarises the evolution, points of technique, results of clinical trials and ongoing research concerning this procedure. Over 3 years of experience with holmium resection/enucleation in two institutions is reviewed. Several
Mohammed S Elsheemy et al.
World journal of urology, 32(4), 985-990 (2013-08-28)
To evaluate the impact of age, stone size, location, radiolucency, extraction of stone fragments, size of ureteroscope and presence and degree of hydronephrosis on the efficacy and safety of holmium:YAG (Ho:YAG) laser lithotripsy in the ureteroscopic treatment of ureteral stones

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service