Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF59183545

Zinc

foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000322

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 487 °C)

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 591-835-45

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

25 mm × 1.0 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
Tandra Ghoshal et al.
Journal of nanoscience and nanotechnology, 9(9), 5586-5591 (2009-11-26)
Nanowire arrays of zinc oxide were synthesized on zinc foil by a simple thermal evaporation process. Morphologies and sizes of the synthesized nanostructures were varied by varying the reaction time and the surface roughness of the substrate. Self-catalytic liquid-solid mechanism
Xufeng Wu et al.
Chemical communications (Cambridge, England), (15)(15), 1655-1657 (2006-04-04)
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).
S Rondot et al.
Science (New York, N.Y.), 263(5154), 1739-1741 (1994-03-25)
An x-ray projection microscope equipped with a charge-coupled device camera allows direct observation of zinc (Zn(2+)) ions diffusing in aqueous hydrochloric acid solution during the corrosion of zinc foil and pellets. Time series of microradiographic images with a lateral resolution
Chuanhui Xia et al.
Journal of nanoscience and nanotechnology, 11(12), 10506-10510 (2012-03-14)
We have investigated the properties of Mn-doped ZnO nanocrystalline film growing on zinc foil by the hydrothermal method. X-ray photoelectron spectroscopy shows that the manganese ions exist as Mn2+ in the film. From UV-vis spectra, we observe a red shift

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service