Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF57563578

Lead

foil, not light tested, 25x25mm, thickness 0.001mm, temporary acrylic support, 99.99+%

Synonym(s):

Lead, PB000020

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 575-635-78

resistivity

20.65 μΩ-cm

size × thickness

25x25 mm × 0.001 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

target_organs

Central nervous system,Blood,Immune system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motoaki Kato et al.
Nihon Hoshasen Gijutsu Gakkai zasshi, 61(8), 1158-1169 (2005-09-01)
In projection radiography, stationary grids are indispensable accessories to the improvement of diagnostic imaging. On the other hand, they are becoming one of the issues facing digital image processing. The lead foil that composes the grid can produce moiré on
M Bourgeois et al.
Health physics, 62(6), 546-552 (1992-06-01)
A significant amount of radiation escapes the rear of an image receptor in conventional dental radiographic procedures despite the placement of a lead-containing backing at the rear of the film pack. The purpose of this investigation was to place additional
Lead foil in the intraoral periapical radiograph as an inclusion agent: a simple method in denture identification.
Thorakkal Shamim
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP, 22(2), 130-131 (2012-02-09)
Lizhi Zhang et al.
Small (Weinheim an der Bergstrasse, Germany), 1(3), 349-354 (2006-12-29)
In this study we demonstrate that one-dimensional (1D) nanostructured lead telluride (PbTe) can be synthesized in a hydrothermal reaction between lead foil and tellurium powder. The resulting materials were characterized by X-ray diffraction, scanning electron microscopy, and transmission electron microscopy.
Tomas Kron et al.
Physics in medicine and biology, 47(5), 713-722 (2002-04-05)
Radioactive sources in close contact offer an alternative to superficial radiation in the treatment of skin lesions. A flattening filter was designed for a lead surface applicator to improve the skin dose distribution of a high dose rate (HDR) brachytherapy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service