Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF54954096

Neodymium

wire reel, 100mm, diameter 1.0mm, hard, 99.9%

Synonym(s):

Neodymium, ND005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 549-540-96

resistivity

64.0 μΩ-cm, 20°C

L × diam.

100 mm × 1.0 mm

bp

3074 °C (lit.)

mp

1021 °C (lit.)

density

7.003 g/mL at 25 °C (lit.)

SMILES string

[Nd]

InChI

1S/Nd

InChI key

QEFYFXOXNSNQGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B S Stein
Mayo Clinic proceedings, 73(8), 787-791 (1998-08-14)
Since 1986, benign prostatic hyperplasia has been treated with lasers, but clinical use was not practical until the right-angled fiber was developed in the early 1990s. The neodymium:yttrium-aluminum-garnet (Nd:YAG) laser is one of four types available for treating the prostate.
B C Patel et al.
Ophthalmology, 104(7), 1191-1197 (1997-07-01)
Laser-assisted dacryocystorhinostomy (DCR) has failed to match the success rates of external DCR. It has been suggested that this technology may be best suited for revision of failed DCR cases. The authors prospectively evaluated the efficacy of transcanalicular laser-assisted revision
J Hellinger
Photomedicine and laser surgery, 22(5), 418-422 (2005-01-27)
In this review, we discuss how nonendoscopic percutaneous laser disc decompression (PLDD) and nucleotomy, using the YAG laser 1064, now has reinforced itself as a minimally invasive procedure in discogenic, vertebral pain syndromes, created by bulging, protrusions, and contained and
J B Dickson et al.
Ophthalmic plastic and reconstructive surgery, 5(1), 17-26 (1989-01-01)
Contact Nd:YAG laser surgery offers a new method of delivering laser energy to a target tissue site. Nd:YAG, a near-infrared laser, can be transmitted through an optical fiber delivery system to a synthetic sapphire probe. This sapphire probe can be
P F Bradley
The British journal of oral & maxillofacial surgery, 35(1), 26-35 (1997-02-01)
A review of the use of neodymium:YAG laser in oral and maxillofacial surgery is presented. The Nd:YAG laser wavelength exhibits minimal tissue absorption and maximal penetration by comparison with the CO2 laser's maximal absorption and minimal penetration. These properties allow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service