Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF53943295

Lead

foil, 15mm disks, thickness 0.007mm, 99.99+%

Synonym(s):

Lead, PB000090

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 539-432-95

resistivity

20.65 μΩ-cm

diam. × thickness

15 mm × 0.007 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

target_organs

Central nervous system,Blood,Immune system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Lopez Medina et al.
Physics in medicine and biology, 49(1), 17-32 (2004-02-20)
Two dosimetry protocols based on absorbed dose to water have recently been implemented: TG-51 and TRS-398. These protocols use different beam-quality indices: %dd(10)x and TPR20,10. The effect of electron contamination in measurements of %dd(10)x has been proposed as a disadvantage
A M Harris et al.
Nuclear medicine communications, 21(9), 851-855 (2000-11-07)
During measurements of the extrinsic spatial resolution of a gamma camera, it was noted that there was a variation with the position in the field of view. The problem was traced to the collimator, and this was confirmed by inspection
Pardeep K Bansal et al.
Contemporary clinical dentistry, 2(2), 76-78 (2011-10-01)
The need for denture labeling is important for forensic and social reasons in case patients need to be identified individually. The importance of denture marking has long been acknowledged by the dental profession. Over the years, various denture marking systems
Leonard J S Tsuji et al.
Journal (Canadian Dental Association), 71(1), 35-38 (2005-01-15)
The lead content of the foil backing of 4 types of intraoral film commonly used by dentists was 69% to 85%. An environmental issue exists because these foils are typically thrown out with regular refuse, even though recycling programs exist.
Steven D Fallon et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 64(2), 223-229 (2006-01-18)
The purpose of this study was to determine if condylar morphology can be accurately depicted on the panoramic radiograph and if variations in condylar anatomy affect the radiographic appearance of the condyle. The shape and angulation of the condyle were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service