GF53461313
Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn
foil, 15mm disks, 0.0125mm thickness, as rolled
Synonym(s):
CO010240
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
form
foil (disks - as rolled)
manufacturer/tradename
Goodfellow 534-613-13
size × thickness
15 mm × 0.0125 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service