Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF52098600

Polyethersulfone

(PES), rod, diam. 55 mm, L 200 mm, dark amber

Synonym(s):

SU307955

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23
Product GF52098600 is not currently sold in your country. Contact Technical Service

form

rod

manufacturer/tradename

Goodfellow 520-986-00

L

200 mm

diam.

55 mm

color

dark amber

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service