GF49999516
Ytterbium
foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Synonym(s):
Ytterbium, YB000230
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
99%
form
foil
manufacturer/tradename
Goodfellow 499-995-16
resistivity
28 μΩ-cm, 20°C
size × thickness
25 x 25 mm × 0.25 mm
bp
1196 °C (lit.)
mp
824 °C (lit.)
density
6.54 g/mL at 25 °C (lit.)
SMILES string
[Yb]
InChI
1S/Yb
InChI key
NAWDYIZEMPQZHO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sethuraman Gayathri et al.
Carbohydrate polymers, 121, 302-308 (2015-02-11)
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region.
Ying Xie et al.
Journal of nanoscience and nanotechnology, 14(6), 4486-4489 (2014-04-18)
Eu3+ and Yb3+ codoped Y2O3 phosphors were synthesized by the sol-gel method. The phosphors possess absorption in the region of 300-550 nm, exhibiting an intense NIR emission of Yb3+ around 1000 nm, which is suitable for matching the maximum spectral
Pr3+/Yb3+ co-doped beta-phase NaYF4 microprisms: controlled synthesis and upconversion luminescence.
Wei Gao et al.
Journal of nanoscience and nanotechnology, 14(6), 4308-4312 (2014-04-18)
Pr3+/Yb3+ co-doped hexagonal NaYF4(beta-NaYF4) microprisms were synthesized by the hydrothermal method, and ethylenediaminetetraacetic acid (EDTA) was introduced to control the size of the microcrystal samples. Bright upconverted fluorescence emission was observed when the samples were excited with an infrared (IR)
Ru Zhou et al.
Journal of nanoscience and nanotechnology, 14(6), 4196-4200 (2014-04-18)
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission
Wanyue Shao et al.
Journal of nanoscience and nanotechnology, 14(5), 3887-3892 (2014-04-17)
Water-soluble BaYF5:Yb3+/Er3+ nanoparticles with the surface functionalized by a layer of poly(acrylic acid) (PAA) were synthesized via a facile one-step PAA-assisted hydrothermal method. Bovine serum albumin (BSA) protein was conjugated with BaYF5:Yb3+/Er3+ upconversion nanoparticles via free carboxylic acid groups on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service