Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF48891976

Polybutylene Terephthalate

(PBT), sheet, thickness 2 mm, size 60 × 60 mm

Synonym(s):

ES343150

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100

form

sheet

manufacturer/tradename

Goodfellow 488-919-76

size

60 × 60 mm

thickness

2 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service