Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF47415045

Aluminum

microfoil, disks, 50mm, thinness 0.25μm, specific density 68.8μg/cm2, permanent mylar 3.5μm support, 99.999%

Synonym(s):

Aluminum, AL004600

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.999%

form

foil

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 474-150-45

resistivity

2.6548 μΩ-cm

L × diam.

50 mm × 0.25 μm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Shaw et al.
Immunologic research, 56(2-3), 304-316 (2013-04-24)
We have examined the neurotoxicity of aluminum in humans and animals under various conditions, following different routes of administration, and provide an overview of the various associated disease states. The literature demonstrates clearly negative impacts of aluminum on the nervous
Maire E Percy et al.
Journal of inorganic biochemistry, 105(11), 1505-1512 (2011-11-22)
In 1991, treatment with low dose intramuscular desferrioxamine (DFO), a trivalent chelator that can remove excessive iron and/or aluminum from the body, was reported to slow the progression of Alzheimer's disease (AD) by a factor of two. Twenty years later
Christopher Auger et al.
Biotechnology advances, 31(2), 266-273 (2012-12-04)
As our reliance on aluminum (Al) increases, so too does its presence in the environment and living systems. Although generally recognized as safe, its interactions with most living systems have been nefarious. This review presents an overview of the noxious
J R Walton
Journal of Alzheimer's disease : JAD, 35(1), 7-43 (2013-02-06)
The neuroanatomic specificity with which Alzheimer's disease (AD) progresses could provide clues to AD etiopathology. Magnetic resonance imaging studies of AD clinical progression have confirmed general conclusions from earlier studies of AD neuropathological progression wherein neurofibrillary tangle pathology was observed
Joseph Lemire et al.
Journal of inorganic biochemistry, 105(11), 1513-1517 (2011-11-22)
Aluminum (Al) has been implicated in a variety of neurological diseases. However, the molecular mechanisms that enable Al to be involved in these disorders have yet to be fully delineated. Using astrocytes as a model of the cerebral cellular system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service