Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF47043378

Tantalum

powder, max. particle size 350 micron, weight 30 g, min. particle size 45 micron

Synonym(s):

TA006010, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

form

powder

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 470-433-78

resistivity

13.5 μΩ-cm, 20°C

weight

30 g

max. part. size

350 μm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service