Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF46902292

Gold

insulated wire, 0.5m, conductor diameter 0.10mm, insulation thickness 0.013mm, PTFE (polytetrafluoroethylene) insulation, 99.99%

Synonym(s):

Gold, AU005837

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 469-022-92

resistivity

2.05 μΩ-cm, 0°C

L × diam. × thickness

0.5 m × 0.10 mm × 0.013 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingbo Zu et al.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Yuanyuan Yao et al.
Journal of nanoscience and nanotechnology, 14(7), 4851-4857 (2014-04-25)
The current study involves in identification and molecular levels characterization of optimal size and concentration of gold nanoparticles (AuNPs). Stable, gold nanoparticles were synthesized and characterized using transmission electron microscopy (TEM) and dynamic light scattering (DLS). The concentration and size
Masaki Ujihara et al.
Journal of nanoscience and nanotechnology, 14(7), 4906-4910 (2014-04-25)
Effect of structure and size of Au nanoparticles (AuNPs) on fluorescence behavior of uranine was examined. Confeito-like AuNPs with different sizes (30 nm, 60 nm and 100 nm, respectively) had plasmon absorption bands at 555, 600 and 660 nm, while
Jiajing Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service