Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF45076822

Moonstone (multicolor)

powder, max. particle size 100 micron, weight 20 g, condition gemstone powder

Synonym(s):

SI856010

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$512.00

$512.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$512.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$512.00


Check Cart for Availability

Request a Bulk Order

form

powder

manufacturer/tradename

Goodfellow 450-768-22

weight

20 g

max. part. size

100 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service