Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF42101413

Brass alloy, Cu63Zn37

foil, 75 x 75mm, 3.0mm thickness, half hard

Synonym(s):

CU020620

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

foil (temper half hard)

manufacturer/tradename

Goodfellow 421-014-13

size × thickness

75 mm × 75 mm × 3.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service