Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF40726829

Nickel-chromium alloy, Ni80Cr20

foil, 6mm disks, 0.25mm thickness, as rolled

Synonym(s):

NI050280

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

foil (disks - as rolled)

manufacturer/tradename

Goodfellow 407-268-29

size × thickness

6 mm × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service