Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF39809863

Holmium

wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%

Synonym(s):

Holmium, HO005105

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 398-098-63

L × diam.

100 mm × 0.5 mm

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Tooher et al.
The Journal of urology, 171(5), 1773-1781 (2004-04-13)
We undertook a systematic review to assess the safety and efficacy of holmium laser prostatectomy compared to transurethral resection of the prostate. We searched literature databases through August 2002. Holmium laser studies, including holmium laser resection of the prostate (HoLRP)
J N Kabalin et al.
Mayo Clinic proceedings, 73(8), 792-797 (1998-08-14)
As alternatives to transurethral resection of the prostate for treatment of benign prostatic hyperplasia, various types of laser prostatectomy-most notably, neodymium:yttrium-aluminum-garnet (YAG) and potassium titanyl phosphate laser procedures-have proved to be associated with considerably less morbidity (for example, bleeding, need
J N Kabalin et al.
Journal of clinical laser medicine & surgery, 16(1), 21-27 (1998-09-05)
The authors review the current knowledge regarding the application of the Holmium: YAG laser for prostatectomy. Conventional surgical therapies for benign prostatic hyperplasia (BPH) are effective but associated with relatively high morbidity. Laser prostatectomy, using either Neodymium:YAG or potassium-titanyl-phosphate lasers
Ramsay L Kuo et al.
Urology, 62(1), 59-63 (2003-07-03)
To review the complications associated with 206 holmium laser enucleation of the prostate (HoLEP) procedures. HoLEP is a minimally invasive surgical treatment for benign prostatic hyperplasia. A retrospective review was conducted of HoLEPs performed from April 1, 1999 to October
T A Wollin et al.
Journal of clinical laser medicine & surgery, 16(1), 13-20 (1998-09-05)
To review the physics related to the holmium laser, its laser-tissue interactions, and its application to the treatment of urological diseases. The holmium: YAG laser is a solid-state, pulsed laser that emits light at 2100 nm. It combines the qualities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service